Learn More
Electron Microscopy Sciences 40-Slide Holder Block, 4"(W) x 12"(L) (10 x 30.5cm)
SDP
Supplier:  Electron Microscopy Sciences 7153240
Encompass_Preferred
Made from solid high density polypropylene blocks with 13 slanted channels (on a 10o angle), 1/16"(1.5 mm) (W) x 3/8" (9 mm) (D). It is made to hold 3x1" (75x25 mm) slides during cleaning, draining excess immersion oil, and even drying. They are white in color and are chemically resistant.
Catalog No. 50-189-9628