Learn More
S2 Media 400mL Synthetic Agar, AOAC, 500mL PC Media Bottle, 1/each
Supplier:  S2 Media 5521
Encompass_Preferred
Synthetic Agar, AOAC is a selective, non-differential, chemically defined medium formulated for the maintenance and propagation of disinfectant test cultures. It supplies all essential nutrients required to sustain the growth of standardized test organisms employed in the determination of phenol coefficients for disinfectant efficacy evaluation.• Also known as Mundy and Wright Agar• Product meets AOAC reference methods and performance criteria.• Synthetic Agar, AOAC should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. NC3900343