Learn More
Sino Biological Influenza A H1N1 (A/Beijing/262/1995) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40133V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Beijing/262/1995(H1N1)) hemagglutinin (ACF41867.1) (Met1-Gln528), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0419