Learn More
Sino Biological Dengue virus DENV-2 (strain New Guinea C) NS5 (methyltransferase domain) / Nonstructural protein 5 Protein (His Tag)
Supplier:  Sino Biological 40267V07E1100
Encompass_Preferred
A DNA sequence encoding the Dengue virus (type 2, strain New Guinea C) NS5 (AAC59275.1) (Gly2492-Gln2787) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-0682