Learn More
Sino Biological Ebola virus EBOV (subtype Bundibugyo, strain Uganda 2007) VP24 Protein (His Tag)
Supplier:  Sino Biological 40456V07E100
Encompass_Preferred
A DNA sequence encoding the Bundibugyo ebolavirus(strain Uganda 2007) VP24 (YP_003815439.1) (Met1-Lys233) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-0907