Learn More
Sino Biological Dromedary camel coronavirus (DcCoV)(strain HKU23-368F) Nucleoprotein / DcCoV-NP Protein (His Tag)
Supplier:  Sino Biological 40458V08B100
Encompass_Preferred
A DNA sequence encoding the Dromedary camel coronavirus (DcCoV)(strain HKU23-368F) NP (AHN64796.1) (Met1-Ile448) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0909