Learn More
Sino Biological Enterovirus D68 (EV-D68) (strain Fermon) VP4 Protein (Fc Tag)
Supplier:  Sino Biological 40467V31B100
Encompass_Preferred
A DNA sequence encoding the VP4 of Enterovirus D68 (EV-D68)(strain Fermon) (AAR98503.1) (Met1-Lys69) was expressed with the Fc region of rabbit IgG at the C-terminus.
Catalog No. 50-162-0940