Learn More
Sino Biological West Nile Virus (WNV) (lineage 2, strain Nea Santa-Greece-2010) E / Envelope Protein (Domain III, His Tag)
Supplier:  Sino Biological 40534V08Y1100
Encompass_Preferred
A DNA sequence encoding the envelope protein of West Nile Virus (WNV) (lineage 2, strain Nea Santa-Greece-2010) (AED99787.1 ) (Gln586-Ala705) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1069