Learn More
New Jersey Fluid Systems LLC Stainless Steel 1-Piece 40G Series Ball Valve, 1.2 Cv, 1/4 in. MNPT
Supplier:  New Jersey Fluid Systems LLC SS43GM4
Encompass
The 40 series one-piece instrumentation ball valve has performed reliably for nearly 40 years in multiple industries. Swagelok modified the valve construction and developed an innovative assembly process. The result is the 40G series ball valve, which features an expanded operating temperature rating, improved thermal cycling capabilities, and reduced potential for leaks.
Catalog No. NC1924760