Learn More
Environmental Sampling Supply Inc 40ML CLEAR OPEN-TOP/.5 ML
Supplier:  Environmental Sampling Supply Inc 40500300QC
Encompass_Preferred
40ML CLEAR BOROSILICATE GLASS VIAL/24-414POLY OPENTOP CLOSURE/TEFLON SILICONE SEPTA 0.5ML HCL EPA CERTIFIED
Catalog No. NC0564713