Learn More
Chemglass Life Sciences Fritted Disc, 40mm, Medium
SDP
Supplier:  Chemglass Life Sciences CGB20130
Encompass_Preferred
Fritted Disc, 40mm, Medium. Porous discs for use in fabrication of special glass apparatus. Fritted glassware is designed for low pressure vacuum filtration or gas flow. The MAX differential pressure should not exceed 15 PSI. Fritted glassware should not be exposed to excessive temperature change or direct exposure to flame due to the fact that it has less resistance to thermal shock than standard borosilicate glassware.
Catalog No. 50-153-1979