Learn More
Chemglass Life Sciences 40mm Quartz Fritted Disc, Coarse
SDP
Supplier:  Chemglass Life Sciences CGQ020720
Encompass_Preferred
40mm Quartz Fritted Disc, Coarse. Porous fused quartz discs. Porosity sizes are as listed and differ from porosities of borosilicate discs.
Catalog No. CGQ020720