Learn More
Uline 40 x 48 Plastic Corrugated Pads, Black
Supplier:  Uline S11311BL
Encompass_Preferred
Reusable plastic corrugated pads provide superior protection, Resists dirt grease and water, Rigid and durable for long-term use, 3/16 thick polypropylene, Pads are corona treated, Corrugation runs lengthwise, Melting Point 160 - 165 F
Catalog No. NC2340922