Learn More
eMolecules 42074-68-0 | 2-Chlorotrityl chloride resin (0.3 - 0.8 mmol/g, 200 - 400 mesh) | Chem-Impex | MFCD00040399 | 313.220 | C19H14Cl2 | 0.000 | Clc1ccccc1C(Cl)(c1ccccc1)c1ccccc1 | 25g | 786483867
Supplier: eMolecules 786483867
2-Chlorotrityl chloride resin (0.3 - 0.8 mmol/g, 200 - 400 mesh) | Chem-Impex | 42074-68-0 | MFCD00040399 | 313.220 | C19H14Cl2 | 0.000 | Clc1ccccc1C(Cl)(c1ccccc1)c1ccccc1 | 25g | 786483867