Learn More
Cary Company 43mm 75 x 35 Clear Perforated Shrink Band
SDP
Supplier:  Cary Company 68WSS4
Encompass
This item has a minimum qty of 3 per supplier requirements.
43mm 75 x 35 Clear Perforated Shrink Band. 43mm 75 x 35 clear perforated shrink band for spice caps. These shrink bands are a cost-effective way to provide tamper-evident protection to packaged goods, from your warehouse location to store shelves. They help provide reassurance for your customers that their spice bottle has not been tampered with. Shrink bands are a cost-effective way to provide tamper-evident protection to packaged goods. Heat shrink bands are manually applied over and around the cap of a container. Heat is applied using a heat gun or heat tunnel until the band shrinks and closely fits around the container.
Catalog No. NC2941652