Learn More
Sigma Aldrich Fine Chemicals Biosciences 44 MethylenebisNN dimethyl500G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences M44451500G
Encompass
4 4AE-Methylenebis(N N-dimethylaniline) is an intermediate in dye manufacture and works as a reagent for the determination of lead. It is suitable for the qualitative assay of cyanogens.
Catalog No. 50-290-7238