Learn More
Chemglass Life Sciences 45/50 Outer PTFE Insert Assembly - PTFE insert with nut, PTFE washer, and perfluoro O-ring; for 45/50 outer joints and reaction vessel lids.
SDP
Supplier:  Chemglass Life Sciences CG1968P130
Encompass_Preferred
PTFE insert assembly for 45/50 outer joints, supplied complete with a nut, PTFE washer, and a perfluoro O-ring. Designed to provide a chemical-resistant seal for reaction vessel lids, multi-neck glassware, and adapters where a 45/50 outer insert is required.
  • Provides a chemical-resistant sealing interface.
  • Supplied complete with a nut, PTFE washer, and perfluoro O-ring.
  • Compatible with 45/50 outer joints and reaction vessel lids.
  • Simple installation for replacement or maintenance.
  • Sold individually (each).
Catalog No. 50-273-3549