Learn More
Chemglass Life Sciences 45/50 Inner Rodavissr Joint, With Drip
SDP
Supplier:  Chemglass Life Sciences CG17408
Encompass_Preferred
45/50 Inner Rodaviss Joint, With Drip. Similar to CG-170, but with a drip tip cut at a 45¿ angle.
Catalog No. CG17408