Learn More
Chemglass Life Sciences Chemglass Quick Connect Set, 1/2 in, 316 Stainless Steel, M16 female, for reactor systems and hoses
Supplier: Chemglass Life Sciences CG1969Q76
The Chemglass quick connect set provides a convenient, durable coupling solution for reactor systems and laboratory hoses. Constructed from 316 stainless steel with fluorosilicone seals, the set features shutoff couplers for safer disconnection and is suitable for a wide temperature range.
- Material: 316 stainless steel.
- Size: 1/2 in.
- Thread type: both ends terminate in M16 female.
- Seals: fluorosilicone.
- Temperature range: -60 to 200 °C.
- Shutoff: both male and female couplers have shutoffs.
- Not dripless/dry: not considered dripless/dry quick disconnect couplings.