Learn More
ABclonal Technology 450 anti-Human CD79b/IgB
SDP
Supplier:  ABclonal Technology A26467500T
Encompass_Preferred
The B lymphocyte antigen receptor is a multimeric complex that includes the antigen-specific component surface immunoglobulin (Ig) Surface Ig non-covalently associates with two other proteins Ig-alpha and Ig-beta which are necessary for expression and function of the B-cell antigen receptor This gene encodes the Ig-beta protein of the B-cell antigen component Alternatively spliced transcript variants encoding different isoforms have been described
Catalog No. 50-000-08997