Learn More
Abcam 450 nm Stop Solution for TMB Substrate, 1000ML
Supplier:  Abcam AB1715291000ML
Encompass_Preferred
Abcam's 450 nm Stop Solution for TMB Substrate is a ready-to-use solution for colorimetric HRP-based ELISA detection.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2261154