Learn More
DAN KAR CORP 48 PRONG FROGGER
Supplier:  DAN KAR CORP MC48
Encompass
Inoculating Manifolds or Froggers, Multiversatile Lab Tool, Replicate transfers between Microtiter Plates are Simple, Used to transfer Cells, make dot Blocks, Grind up Drosophila in Microtiter Plates and more, Aluminum base, Autoclavable
Catalog No. NC9843368