Learn More
Chemglass Life Sciences Chemglass 48-Well Flat Bottom Tissue Culture Plate, TC-treated, Sterile, 100% Virgin Polystyrene
SDP
Supplier:  Chemglass Life Sciences CGN3500B48
Encompass_Preferred
48-well flat bottom tissue culture plate made from high-clarity, 100% virgin polystyrene. The plate is tissue culture treated by vacuum plasma to promote cell attachment, sterilized by E-beam, and designed with a low-evaporation lid and small feet to protect microscope objectives. Recommended medium volume per well is 0.19-0.285 mL, with a cell growth area of 0.88 cm².
  • 48 wells, flat bottom suitable for microscopy
  • Made from high-clarity, 100% virgin polystyrene
  • Tissue culture treated by vacuum plasma for improved cell adhesion
  • E-beam sterilized and DNase/RNase-free
  • Low-evaporation lid design for uniform growth
  • Recommended medium volume: 0.19-0.285 mL per well
  • Cell growth area per well: 0.88 cm²
  • Individually wrapped packaging for aseptic handling
  • Clear lot number for batch traceability
Catalog No. 50-273-3438