Learn More
Analytical Sales & Services Analytical Sales 48-Position TCR, Precise Thermal Management, Enhanced Reproducibility, High Throughput
Supplier:  Analytical Sales & Services 489900
Encompass
The Analytical Sales & Services 48-Position Temperature Controlled Reactor is designed for precise thermal management in high-throughput experimentation, ensuring uniform temperature control and enhanced reproducibility. Its versatility allows compatibility with various heat transfer fluids and laboratory automation, making it ideal for demanding chemical and pharmaceutical applications. The reactor supports photoredox configurations with additional components, setting it apart from standard models.
Catalog No. NC2172898