Learn More
ABclonal Technology 488 anti-Human CD96/TACTILE
SDP
Supplier:  ABclonal Technology A24277200T
Encompass_Preferred
The protein encoded by this gene belongs to the immunoglobulin superfamily It is a type I membrane protein The protein may play a role in the adhesive interactions of activated T and NK cells during the late phase of the immune response It may also function in antigen presentation Alternative splicing generates multiple transcript variants encoding distinct isoforms
Catalog No. 50-000-08954