Learn More
ABclonal Technology 488 Rabbit anti- CD162/PSGL-1
SDP
Supplier:  ABclonal Technology A25940100T
Encompass_Preferred
Involved in leukocyte adhesive activation Acts upstream of or within leukocyte tethering or rolling Predicted to be located in plasma membrane raft and uropod Predicted to be active in plasma membrane Is expressed in brain and thymus primordium Human ortholog(s) of this gene implicated in carotid artery disease Orthologous to human SELPLG (selectin P ligand)
Catalog No. 50-000-10806