Learn More
ABclonal Technology 488 Rabbit anti-Human CD24
SDP
Supplier:  ABclonal Technology A25380200T
Encompass_Preferred
This gene encodes a sialoglycoprotein that is expressed on mature granulocytes and B cells and modulates growth and differentiation signals to these cells The precursor protein is cleaved to a short 32 amino acid mature peptide which is anchored via a glycosyl phosphatidylinositol (GPI) link to the cell surface This gene was missing from previous genome assemblies but is properly located on chromosome 6 Non-transcribed pseudogenes have been designated on chromosomes 1 15 20 and Y Alternative splicing results in multiple transcript variants
Catalog No. 50-000-12906