Learn More
ABclonal Technology 488 Rabbit anti-Human CD35
SDP
Supplier:  ABclonal Technology A24697100T
Encompass_Preferred
Complement receptor type 1 (CR1 CD35) is a type I transmembrane glycoprotein that is expressed on the surface of B cells neutrophils monocytes and renal podocytes As a component of the host s innate immune system CR1 CD35 expressed on neutrophils and monocytes binds to ligands coated with complement opsonins C3b and C4b thereby promoting phagocytosis and pro-inflammatory cytokine production CR1 CD35 also participates in negative regulation of the complement cascade through its ability to promote dissociation of C3 and C5 convertases and by serving as one of multiple cofactors for factor-I-mediated cleavage and inactivation of C3b and C4b
Catalog No. 50-000-09902