Learn More
ABclonal Technology 488 Rabbit anti-Human CD52
SDP
Supplier:  ABclonal Technology A26721100T
Encompass_Preferred
Involved in positive regulation of cytosolic calcium ion concentration Predicted to be located in extracellular region and plasma membrane Predicted to be intrinsic component of plasma membrane Predicted to be active in sperm midpiece
Catalog No. 50-000-11196