Learn More
ABclonal Technology 488 Rabbit anti-Human CD71
SDP
Supplier:  ABclonal Technology A22301500T
Encompass_Preferred
This gene encodes a cell surface receptor necessary for cellular iron uptake by the process of receptor-mediated endocytosis This receptor is required for erythropoiesis and neurologic development Multiple alternatively spliced variants have been identified
Catalog No. 50-000-07184