Learn More
ABclonal Technology 488 Rabbit anti-Human CD74
SDP
Supplier:  ABclonal Technology A26657100T
Encompass_Preferred
The protein encoded by this gene associates with class II major histocompatibility complex (MHC) and is an important chaperone that regulates antigen presentation for immune response It also serves as cell surface receptor for the cytokine macrophage migration inhibitory factor (MIF) which when bound to the encoded protein initiates survival pathways and cell proliferation This protein also interacts with amyloid precursor protein (APP) and suppresses the production of amyloid beta (Abeta) Multiple alternatively spliced transcript variants encoding different isoforms have been identified
Catalog No. 50-000-11032