Learn More
ABclonal Technology 488 Rabbit anti-Human CR2/CD21
SDP
Supplier:  ABclonal Technology A22643100T
Encompass_Preferred
This gene encodes a membrane protein which functions as a receptor for Epstein-Barr virus (EBV) binding on B and T lymphocytes Genetic variations in this gene are associated with susceptibility to systemic lupus erythematosus type 9 (SLEB9) Alternatively spliced transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-05154