Learn More
ABclonal Technology 488 Rabbit anti-Human FAP
SDP
Supplier:  ABclonal Technology A24965500T
Encompass_Preferred
The protein encoded by this gene is a homodimeric integral membrane gelatinase belonging to the serine protease family It is selectively expressed in reactive stromal fibroblasts of epithelial cancers granulation tissue of healing wounds and malignant cells of bone and soft tissue sarcomas This protein is thought to be involved in the control of fibroblast growth or epithelial-mesenchymal interactions during development tissue repair and epithelial carcinogenesis Alternatively spliced transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-12256