Learn More
ABclonal Technology 488 Rabbit anti-Human GSK3B
SDP
Supplier:  ABclonal Technology A26725100T
Encompass_Preferred
The protein encoded by this gene is a serine-threonine kinase belonging to the glycogen synthase kinase subfamily It is a negative regulator of glucose homeostasis and is involved in energy metabolism inflammation ER-stress mitochondrial dysfunction and apoptotic pathways Defects in this gene have been associated with Parkinson disease and Alzheimer disease
Catalog No. 50-000-12154