Learn More
ABclonal Technology 488 Rabbit anti-Human P4HB
SDP
Supplier:  ABclonal Technology A26906200T
Encompass_Preferred
This gene encodes the beta subunit of prolyl 4-hydroxylase a highly abundant multifunctional enzyme that belongs to the protein disulfide isomerase family When present as a tetramer consisting of two alpha and two beta subunits this enzyme is involved in hydroxylation of prolyl residues in preprocollagen This enzyme is also a disulfide isomerase containing two thioredoxin domains that catalyze the formation breakage and rearrangement of disulfide bonds Other known functions include its ability to act as a chaperone that inhibits aggregation of misfolded proteins in a concentration-dependent manner its ability to bind thyroid hormone its role in both the influx and efflux of S-nitrosothiol-bound nitric oxide and its function as a subunit of the microsomal triglyceride transfer protein complex
Catalog No. 50-000-08573