Learn More
ABclonal Technology 488 Rabbit anti-Human PLZF
SDP
Supplier:  ABclonal Technology A27688500T
Encompass_Preferred
This gene is a member of the Krueppel C2H2-type zinc-finger protein family and encodes a zinc finger transcription factor that contains nine Kruppel-type zinc finger domains at the carboxyl terminus This protein is located in the nucleus is involved in cell cycle progression and interacts with a histone deacetylase Specific instances of aberrant gene rearrangement at this locus have been associated with acute promyelocytic leukemia (APL) Alternate transcriptional splice variants have been characterized
Catalog No. 50-000-10085