Learn More
ABclonal Technology 488 Rabbit anti-Human PTH1R
SDP
Supplier:  ABclonal Technology A26912200T
Encompass_Preferred
The protein encoded by this gene is a member of the G-protein coupled receptor family 2 This protein is a receptor for parathyroid hormone (PTH) and for parathyroid hormone-like hormone (PTHLH) The activity of this receptor is mediated by G proteins which activate adenylyl cyclase and also a phosphatidylinositol-calcium second messenger system Defects in this receptor are known to be the cause of Jansen s metaphyseal chondrodysplasia (JMC) chondrodysplasia Blomstrand type (BOCD) as well as enchodromatosis Two transcript variants encoding the same protein have been found for this gene
Catalog No. 50-000-08491