Learn More
Aviva Systems Biology Corp 4930572J05Rik Antibody - C-terminal region (ARP47289_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47289_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against 4930572J05Rik. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Mouse 4930572J05Rik. Alternate names: MGC54796, 4930572J05RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-285-295