Learn More
Medchemexpress LLC 4A3-Cit | 2768211-13-6 | C83H147N3O16S4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1533735MG
Encompass_Preferred
4A3-Cit is an ionizable lipid used for the generation of lipid nanoparticles (LNPs). It facilitates endosomal escape by undergoing lipid fusion with endosomal membranes within the model.
  • Enhances the efficiency of in vivo mRNA delivery through selective organ targeting (SORT)
Catalog No. 50-003-73377