Learn More
Cayman Chemical 4amIno1 8NaphthalImIde 100mg
SDP
Supplier:  Cayman Chemical 18509100 MG
Encompass_Preferred
An inhibitor of PARP (IC50 = 180 nM); blocks radiation-induced PARP in cancer cells, potentiating the cytotoxicity of γ-radiation, although it is not cytotoxic in the absence of radiation
Catalog No. 50-296-8030