Learn More
Cayman Chemical 4AmInobenzoIc AcIdodIum s 250g
SDP
Supplier:  Cayman Chemical 18659250 G
Encompass_Preferred
An intermediate in the synthesis of tetrahydrofolic acid in many non-mammalian organisms, including bacteria and fungi; metabolized by a variety of enzymes in mammals, including N-acetyltransferases; may also be utilized by bacteria or fungi that are living in mammalian organisms
Catalog No. 50-296-6015