Learn More
Cayman Chemical 4Carboxybutyld4trIphenylp 5mg
SDP
Supplier:  Cayman Chemical 312545 MG
Encompass_Preferred
An internal standard for the quantification of (4-carboxybutyl)triphenylphosphonium by GC- or LC-MS
Catalog No. 50-297-8956