Learn More
Lonza Walkersville 4D-Nucleofector Module Rack
Supplier:  Lonza Walkersville AAK2001
Encompass
The 4D-Nucleofector Module Rack supports more convenient handling of 20 µL 16-well Nucleocuvette Strips.Benefits:Tailor-made rack for easy handling of 16-well Nucleocuvette StripsContent:1 Unit
Catalog No. NC1691511