Learn More
TARGETMOL CHEMICALS INC 4E1RCAT 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T174225MG
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 4E1RCat is a dual inhibitor of eIF4EeIF4 g and eIF4E4E-BP1 interaction. And it inhibits the binding of eIF4 g to eIF4E with IC50 of 3.2 uM. purity: 99%
Catalog No. 50-258-1103