Learn More
Cayman Chemical 4EpInhydrochLRtetracyclIn 5mg
SDP
Supplier:  Cayman Chemical 231465 MG
Encompass_Preferred
An inhibitor of MMP-9; a derivative of tetracycline; inhibits MMP-9 activity in a dose-dependent manner in an assay for collagen degradation; completely inhibits neutrophil-derived MMP-9 degradation of denatured collagen at a concentration of 500 μg/ml; lacks antibiotic activity
Catalog No. 50-299-4060