Learn More
Cayman Chemical 4fluoroaPyrroldInobutIoph 10mg
SDP
Supplier:  Cayman Chemical 1551610 MG
Encompass_Preferred
An analog of 4-MePBP in which a fluorine group is added at the four position of the phenyl ring; intended for forensic and research applications
Catalog No. 50-297-1280