Learn More
AirSea USA 4GV Boxes for Non-specific inners, 4GV/X5/S, external Dimensions: 8.27" x 8.27" x 14.37" certified for Lithium Batteries, Glass, Metal and Plastic inner containers
Supplier:  AirSea USA 838
Encompass_Preferred
4GV Fibreboard box suitable for shipping dangerous goods by road, sea or air inside various inner packagings. UN Mark: 4GV/X5/S. Sold complete with plastic liner bag and suitable for non-specific inner packagings. Vermiculite must be used as a dangerous goods packaging material when shipping items within this 4GV box range.
Catalog No. NC2101123