Learn More
Medchemexpress LLC 4H-1-Benzopyran-4-one, 3-hydroxy-7-methoxy-2-phenyl- | 7478-60-6 | 98.5% | 268.26 | 10 MG

Supplier: Medchemexpress LLC HYN924110MG
3-Hydroxy-7-methoxyflavone is a flavonoid isolated from the natural plant *Salvia elegans*. This compound exhibits anti-anxiety effects and is intended for research use only, as it has not been fully validated for medical applications.
- Flavonoid compound
- Isolated from *Salvia elegans*
- Exhibits anti-anxiety effects
- Appears as a white to off-white solid
- Store at room temperature for 3 years
- Store in solvent at -80°C for 2 years, or -20°C for 1 year
- Soluble in DMSO
- Intended for research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.