Learn More
Medchemexpress LLC 4H-1-benzopyran-4-one, 6-[(1S,5R,6S)-6-(2,4-dihydroxybenzoyl)-5-(2,4-dihydroxyphenyl)-3- | 85698-31-3 | 97.2% | 694.72 g/mol | C40H38O11 | 1MG
SDP
Supplier:  Medchemexpress LLC HY123721MG
Encompass_Preferred
Sanggenon G is a cell-permeable small molecule inhibitor of X-linked inhibitor of apoptosis protein (XIAP) that binds the BIR3 domain and enhances caspase activation (reported binding affinity 34.26 μM). The compound is supplied as a solid (CAS 85698-31-3), has molecular formula C40H38O11 and molecular weight 694.72 g/mol, and is reported at 97.23% purity in the certificate of analysis.
  • Inhibits XIAP by binding the BIR3 domain.
  • Enhances caspase activation in cell-based assays.
  • Characterized by molecular formula C40H38O11 and MW 694.72 g/mol.
  • High reported purity suitable for research applications (97.23%).
  • Available in small milligram quantities for laboratory use.
Catalog No. 50-002-37610