Learn More
Medchemexpress LLC 4h-1-Benzopyran-4-one, 6-[3-(4-morpholinyl)propyl]-2-thieno[3,2-c]pyridin-6-yl-, oxime, hydrochloride | 2133294-96-7 | 98.6% | 457.97 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY108703A
Encompass_Preferred
Foliglurax monohydrochloride is a highly selective and potent, brain-penetrant metabotropic glutamate receptor 4 positive allosteric modulator (mGluR4 PAM) with an EC50 of 79 nM. It exhibits an antiparkinsonian effect and is for research use only.
  • Highly selective and potent mGluR4 PAM.
  • Brain-penetrant.
  • Exhibits an antiparkinsonian effect.
  • For research use only.
  • Potentially revolutionizing mGlu4 receptor drug targeting in CNS disorders.
Catalog No. 50-003-47494